Learn More
Invitrogen™ Second Strand cDNA Synthesis Kit
Invitrogen™ Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.
$1264.00 - $4215.00
Specifications
Product Type | cDNA Synthesis Kit |
---|---|
For Use With (Application) | Cloning |
Final Product Type | Double Stranded cDNA |
Reaction Format | Separate Components |
Description
InvitrogenTM Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.
Features of the Second Strand cDNA Synthesis Kit include:
- Efficient synthesis of full-length double-stranded cDNA
- Fast—procedure completed in 1 hour
- Convenient—pre-mixed components
- Complete—includes residual RNA removal reagents
Applications
- Full-length double-stranded blunt-end cDNA synthesis for cloning
- Double-stranded cDNA library construction
Specifications
cDNA Synthesis Kit | |
Cloning | |
Double Stranded cDNA | |
Separate Components |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.