Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Secretin R Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306386APC
Description
Secretin R Polyclonal antibody specifically detects Secretin R in Human samples. It is validated for Western BlotSpecifications
Secretin R | |
Polyclonal | |
Western Blot | |
pancreatic secretin receptor, SCT-R, secretin receptor, SR | |
Produced in rabbits immunized with E. coli-derived Human Secretin R fragment. | |
0.1 mL | |
GPCR | |
6344 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction