Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Secretin R Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306386IR
This item is not returnable.
View return policy
Description
Secretin R Polyclonal antibody specifically detects Secretin R in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Secretin R | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| DyLight 755 | |
| pancreatic secretin receptor, SCT-R, secretin receptor, SR | |
| Produced in rabbits immunized with E. coli-derived Human Secretin R fragment. | |
| 0.1 mL | |
| GPCR | |
| 6344 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction