Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Secretin R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306386100UL
Description
Secretin R Polyclonal antibody specifically detects Secretin R in Human samples. It is validated for Western BlotSpecifications
Secretin R | |
Polyclonal | |
Western Blot 1:500-1:2000 | |
pancreatic secretin receptor, SCT-R, secretin receptor, SR | |
Produced in rabbits immunized with E. coli-derived Human Secretin R fragment. | |
100 μg | |
GPCR | |
6344 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction