Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Secretory component (free and bound) Mouse anti-Human, Biotin, Clone: NI 194-4 (A89-39), Nordic
SDP

Catalog No. 502100992
Encompass
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0992 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-0992 Supplier Vector Laboratories Supplier No. MAHU/SC/BIO
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Clone 22D3 is an antibody highly specific for a- cardiac actin, and does not cross react with other actin isoforms. The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.

Secretory component (free and bound) Monoclonal antibody specifically detects Secretory component (free and bound) in Human samples. It is validated for Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Secretory component (free and bound)
Applications Dot Blot, ELISA, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NI 194-4 (A89-39)
Concentration 0.4 mg/mL
Conjugate Biotin
Formulation PBS with no preservative; pH
Gene Pigr
Gene Accession No. P01833
Gene Alias hepatocellular carcinoma associated protein TB6; hepatocellular carcinoma-associated protein TB6; phosphatidylinositol glycan, class R; Pigr; Poly Ig receptor; poly-Ig receptor; polymeric immunoglobulin receptor; Secretory component; tb6
Host Species Mouse
Immunogen Human secretory component (free and bound)
Purification Method Protein G
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5284
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.