Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Secretory IgA Monoclonal Antibody (Mc29-12)
GREENER_CHOICE

Catalog No. PIMA181174
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA181174 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA181174 Supplier Invitrogen™ Supplier No. MA181174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This clone recognizes purified human secretory chain and secretory IgA. It shows no reaction against myeloma IgA (native or reduced and alkylated), and is negative against normal human IgM, myeloma IgM, normal human IgG; free kappa or gamma chains, and J-chain. This antibody is recommended for frozen and paraffin embedded sections of intestinal and breast tissue.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Secretory IgA
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Mc29-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with <0.1% sodium azide
Gene Alias Human IgA
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.