Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SECTM1 Antibody (01), Alexa Fluor™ 405 , Novus Biologicals™
SDP

Catalog No. NB223751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB223751 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB223751 Supplier Novus Biologicals Supplier No. NBP305807AF405
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SECTM1 Monoclonal antibody specifically detects SECTM1 in Human samples. It is validated for ELISA

Specifications

Antigen SECTM1
Applications ELISA
Classification Monoclonal
Clone 01
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SECTM1 (Q8WVN6-1; Met1-Gly145).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6398
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.