Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECTM1 Antibody (01), FITC , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305807F
Description
SECTM1 Monoclonal antibody specifically detects SECTM1 in Human samples. It is validated for ELISASpecifications
SECTM1 | |
Monoclonal | |
FITC | |
K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SECTM1 (Q8WVN6-1; Met1-Gly145). | |
0.1 mL | |
Cytokine Research | |
6398 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
01 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction