Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECTM1 Mouse anti-Human, Clone: 1C11B4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SECTM1 |
---|---|
Clone | 1C11B4 |
Concentration | 0.83 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6376
![]() ![]() |
Proteintech Group Inc
602811IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to SECTM1.
This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.Specifications
SECTM1 | |
0.83 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
K12, Protein K 12, secreted and transmembrane 1, SECTM1 | |
SECTM1 | |
IgM | |
Caprylic Acid Precipitation | |
Antibody |
1C11B4 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8WVN6 | |
6398 | |
SECTM1 Fusion Protein Ag14645 | |
Primary | |
-20°C | |
SECTM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title