Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SECTM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584056
Description
Immunogen sequence: CRCSQQRREK KFFLLEPQMK VAALRAGAQQ GLSRASAELW TPDSEPTPRP LALVFKPSPL GALELLSPQP LFPYAAD.
This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.
Specifications
SECTM1 | |
Polyclonal | |
Unconjugated | |
SECTM1 | |
1810003C24Rik; K12; Protein K-12; secreted and transmembrane 1; secreted and transmembrane 1B; secreted and transmembrane protein 1; secreted and transmembrane protein 1b; SECTM1; Sectm1b; type 1a transmembrane protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6398 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVN6 | |
SECTM1 | |
Recombinant protein corresponding to Human SECTM1. Recombinant protein control fragment (Product #RP-101333). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction