Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SECTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542725 Supplier Invitrogen™ Supplier No. PA542725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ARDSHAGLYM WHLVGHQRNN RQVTLEVSGA EPQSAPDTGF WPVPAVVTAV Sequence homology: Human: 100%.

This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SECTM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SECTM1
Gene Accession No. Q8WVN6
Gene Alias 1810003C24Rik; K12; Protein K-12; secreted and transmembrane 1; secreted and transmembrane 1B; secreted and transmembrane protein 1; secreted and transmembrane protein 1b; SECTM1; Sectm1b; type 1a transmembrane protein
Gene Symbols SECTM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SECTM1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6398
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.