Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECTM1 Rabbit anti-Human, Clone: 106, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290085100UL
Description
SECTM1 Monoclonal specifically detects SECTM1 in Human samples. It is validated for ELISA.Specifications
| SECTM1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 106 | |
| ELISA 1:5000-1:10000 | |
| K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein | |
| Recombinant Human SECTM1 Protein | |
| 100 μg | |
| Cytokine Research | |
| 6398 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction