Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECTM1 Rabbit anti-Human, Clone: 106, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29008550UL
Description
SECTM1 Monoclonal specifically detects SECTM1 in Human samples. It is validated for ELISA.Specifications
SECTM1 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
106 | |
ELISA 1:5000-1:10000 | |
K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein | |
Recombinant Human SECTM1 Protein | |
50 μg | |
Cytokine Research | |
6398 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction