Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SecurAlert Clip-On Badges

Catalog No. 19103152 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Application):
Contractor
Temporary
Visitor
Item Description:
1 day expiration
1 week expiration
Color:
Blue
Green
Red
White
Yellow
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Item Description Color
19-103-152 Visitor 1 week expiration Yellow
19-103-150 Visitor 1 week expiration Red
19-103-153 Visitor 1 week expiration Green
19-103-154 Visitor 1 week expiration White
19-103-163 Contractor 1 week expiration Green
19-103-165 Temporary 1 day expiration Red
19-103-169 Temporary 1 day expiration White
19-103-170 Temporary 1 week expiration Red
19-103-174 Temporary 1 week expiration White
19-103-166 Temporary 1 day expiration Blue
19-103-167 Temporary 1 day expiration Yellow
19-103-173 Temporary 1 day expiration Green
19-103-151 Visitor 1 week expiration Blue
19-103-161 Contractor 1 week expiration Blue
19-103-162 Contractor 1 week expiration Yellow
19-103-171 Temporary 1 week expiration Blue
19-103-172 Temporary 1 week expiration Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 19-103-152 Supplier Brady™ Supplier No. 95650

Prevent unauthorized use of visitor, contractor and temporary passes. Brady™ SecurAlert Clip-On Badges are self-expiring, which prevents unauthorized use beyond their intended use periods.

  • Self-expiring badges
  • Low cost clip-on alternative to self-adhesive badges
  • 3 in. x 3 in. slotted cards
  • 500 front and back badge pairs per pack

Specifications

Item Description 1 week expiration
Length (English) 3 in.
Color Yellow
Dimensions (L x W) 3 in. x 3 in.
For Use With (Application) Visitor
Width (English) 3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.