Learn More
Description
- Self-expiring badges
- Low cost clip-on alternative to self-adhesive badges
- 3 in. x 3 in. slotted cards
- 500 front and back badge pairs per pack
Specifications
Specifications
| Item Description | 1 week expiration |
| Length (English) | 3 in. |
| Color | Blue |
| Dimensions (L x W) | 3 in. x 3 in. |
| For Use With (Application) | Temporary |
| Width (English) | 3 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.