Learn More
Description
- Low-cost clip-on alternative to self-adhesive badges
- Package contains 500 front and back parts
- Self-expiring cards prevent unauthorized use
Specifications
Specifications
| Length (English) | 3 in. |
| Color | Green |
| Dimensions (L x W) | 3 in. x 3 in. |
| For Use With (Application) | Visitors |
| Includes | Two parts per card |
| Width (English) | 3 in. |
| Product Type | One-day expiration |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.