Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ SeCure Y Connector Kits
SDP

Catalog No. 06611149 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
0.18/0.25/0.28 mm ID Columns
0.45/0.53 mm ID Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
06-611-149 0.45/0.53 mm ID Columns
06-611-148 0.18/0.25/0.28 mm ID Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-149 Supplier Restek™ Supplier No. 20278
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ferrules and Knurled Nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting even at temperatures as high as 400°C

  • Combines the simplicity of a Y Press-Tight connector with the strength of a metal union
  • Connect two analytical columns to a transfer line or guard column
  • Use standard Y Press-Tight connectors and 1/16 in. graphite ferrules
  • Reliable seal integrity will not unexpectedly disconnect during temperature-programmed analyses
  • Open design allows visual confirmation of the seal for added confidence in the connection
  • Fits both Restek cage designs

Specifications

Color Mixed
Includes SeCure Y Connector Body (1), Knurled Nuts (3), Universal Y Press-Tight Union (1), and Ferrules (3)
For Use With (Application) 0.45/0.53 mm ID Columns
Max. Operating Temperature (Metric) 400°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.