Learn More
3M™ SecureFit™ 200 Series Anti-Fog Lens
3M™ SecureFit™ 200 Series Anti-Fog Lens provides users with blue temples and lenses to help make eyewear more visible in food preparation and processing areas.
$170.06
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1
Product Type Protective Eyewear
For Use With (Application) Food and Beverage Manufacturing
Frame Color Blue
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Quantity Price Quantity & Availability  
19-070-940
View Documents
3M™
7100180044
Clear Case of 20
Case of 20 for $170.06
Only null left
Add to Cart
19-070-941
View Documents
3M™
7100180046
Blue Case of 20
Case of 20 for $170.06
Only null left
Add to Cart
Description
Description
  • Blue temples can be more visible if found in food preparation and processing areas
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fitBlue temples can be more visible if found in food preparation and processing areas
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings
  • Molded nose bridge temples can be more visible if found in food preparation and processing areas
  • Blue lenses are helpful to reduce glare, eye stress and fatigue in yellow light environments, such as produced by sodium vapor bulbs
  • 3M Catalog# SF201SGAF-BLU , 3M Catalog# SF204SGAF-BLU
Specifications
Specifications
Anti-fog, Scratch-resistant
Protective Eyewear
Blue
20/Pk.
Polycarbonate
Frameless
ANSI Z87.1
Food and Beverage Manufacturing
Case of 20
Pressure Diffusion Temple Technology
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.