Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Securin / PTTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Securin / PTTG1 |
---|---|
Concentration | 0.38 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1953
![]() ![]() |
Proteintech Group Inc
180401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Pituitary tumor-transforming gene 1 (PTTG-1) is an oncogene expressed in normal tissues containing proliferating cells and in tumors. PTTG is downstream of the MAP kinase signaling cascade and is phosphorylated at Ser162 by MAPK in vitro. Phosphorylation plays an essential role in PTTG′s transactivation function. The PTTG protein contains a DNA-binding domain, and it activates transcription of the c-Myc oncogene by binding to the promoter to form a complex. Studies have demonstrated that expression of human PTTG correlates with cell proliferation in a cell cycle dependent manner, and the association of human PTTG with Ku, the regulatory subunit of DNA-dependent protein kinase, is disrupted when a DNA double-strand breaks occurs.Specifications
Securin / PTTG1 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95997, P97613, Q9CQJ7 | |
30939, 64193, 9232 | |
Securin / PTTG1 Fusion Protein Ag12670 | |
Primary | |
-20°C | |
PTTG1 |
0.38 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EAP1, Esp1 associated protein, HPTTG, pituitary tumor transforming 1, PTTG, PTTG1, Securin, Tumor transforming protein 1, TUTR1 | |
Pttg1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title