Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Securin Recombinant Rabbit Monoclonal Antibody (JB37-37)
GREENER_CHOICE

Catalog No. PIMA541255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541255 Supplier Invitrogen™ Supplier No. MA541255
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Securin Recombinant Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

The encoded protein is a homolog of yeast securin proteins, which prevent separins from promoting sister chromatid separation. It is an anaphase-promoting complex (APC) substrate that associates with a separin until activation of the APC. The gene product has transforming activity in vitro and tumorigenic activity in vivo, and the gene is highly expressed in various tumors. The gene product contains 2 PXXP motifs, which are required for its transforming and tumorigenic activities, as well as for its stimulation of basic fibroblast growth factor expression. It also contains a destruction box (D box) that is required for its degradation by the APC. The acidic C-terminal region of the encoded protein can act as a transactivation domain. The gene product is mainly a cytosolic protein, although it partially localizes in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Securin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB37-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Pttg1
Gene Accession No. O95997
Gene Alias AW555095; C87862; EAP1; Esp1-associated protein; ESP1-associated protein 1; HPTTG; MGC126883; MGC138276; Pituitary tumor transforming; pituitary tumor-transforming 1; pituitary tumor-transforming gene 1; pituitary tumor-transforming gene 1 protein; PTTG; PTTG1; Pttg3; Securin; tumor-transforming protein 1; TUTR1
Gene Symbols Pttg1
Host Species Rabbit
Immunogen Recombinant protein within C-terminal Human Securin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.