Learn More
PDC Securline™ Blood Barcode Wristband, Tail with Barcode and Alpha Numeric Labels
Using bar code and serialized alphanumeric codes, the Securline™ Bar Code Blood Band accurately matches the right patient to the right blood, ensuring maximum safety throughout the blood transfusion process.
$463.00 - $474.00
Specifications
Includes 100/Box
Closure Type Snap
Material Polypropylene, PET
Product Type Blood Band
Dimensions (L x W) 3.25 x 11 in.
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-138-371
View Documents
PDC
243316PDB
Red
Case of 150 for $474.00
Only null left
Add to Cart
22-138-353
View Documents
PDC
243322PDB
Green
Case of 150 for $463.00
Only null left
Add to Cart
Description
Description
  • Wristband labels with large alphanumeric font (14 pt.) and barcodes
  • Total of 20 matching labels (8 alphanumeric and barcode on wristband; 12 barcode/alphanumeric on draw tube tail)
  • Streamlines the blood transfusion process and improves patient safety by reducing human errors related to patient identification
  • Saves time and money by streamlining the patient ID and blood transfusion workflow and minimizes paperwork
  • Integrates seamlessly with your existing blood bank system
  • Patented dual-purpose, hybrid style lets you handwrite information or use with your LIS label
  • Standardize and consolidate to a single blood band, even if you have multiple labs
  • Designed for inpatient or outpatient use
  • Laminate shield protects patient information and bar code from water or solvents for accurate bar code scanning
  • Advanced material resists moisture, alcohol and hand sanitizer to preserve bar code integrity for accurate scanning
  • Meets The Joint Commission's National Patient Safety Goal for blood transfusions for a one-person verification system
Specifications
Specifications
100/Box
Polypropylene, PET
3.25 x 11 in.
Snap
Blood Band
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.