Learn More
Eisco™ Sedimentary Rocks Kit with Storage Box and ID Card
SCI_ED
Catalog No. S012304 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Sedimentary Rock
Catalog No. Product Type
S012304 Sedimentary Rock
1 options
Catalog No. S012304 Supplier Eisco™ Supplier No. ESNGKIT0009
Add to Cart
Edge
Add to Cart
A fun introduction to the world of geology through a 12-piece rock kit. Each Tech Cut specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features.
  • Sedimentary rocks are formed by the deposition and compression of particles of other rocks and minerals
  • Eisco Labs™ Tech Cut Rock and Mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Set includes twelve different sedimentary rocks: Breccia, Bituminous Coal, Conglomerate, Dolostone, Greywacke, Gray Limestone, Limestone Chalk, Fossiliferous Limestone, Coquina, Olitic Limestone, White/Tan Limestone and Red Sandstone
  • While each specimen will vary in shape and size, they will measure approximately 1 in. each
  • Test hardness, streak color, magnetic properties, appearance, and effervescence to identify the different specimens in this kit
  • Packaged in a box with substantial protective foam padding to store and preserve specimens
  • Ideal for home schooling or a fun home science activity, and is a great addition to any classroom or geology laboratory
  • Useful for demonstrations in identifying and classifying rocks and minerals, especially for group activities
Specifications
Product Type Sedimentary Rock