Learn More
Eisco™ Sedimentary Rocks Kit
SCI_ED
Catalog No. S24665 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Specimens
Catalog No. Quantity
S24665 12 Specimens
1 options
Catalog No. S24665 Supplier Eisco™ Supplier No. ESNG0006
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For use in identification of depositional history of sedimentary rocks based on their grain size and composition. The Sedimentary Rocks Kit contains 12 specimens and specimen key.
  • Classroom tool for sedimentary rock classification and identification
  • Numbered and stored in sturdy, sectioned container for easy access and safety
  • Average specimen size: 2 to 3 cm
  • Size and shape of grains can be used to identify what kind of environment would have formed particular rock
Specifications
Product Type Sedimentary Rock Kit
For Use With (Application) Identify depositional history of sedimentary rocks based on their grain size and composition
Includes Bituminous coal, breccia, conglomerate, dolostone, rock gypsum, gray and fossiliferous limestone, red and white sandstone, argillaceous and carbonaceous shale, siltstone
Packaging Sectioned container
Quantity 12 Specimens
Specimen Size Approx. 1 in.