Learn More
Invitrogen™ SeeBlue™ Plus2 Pre-stained Protein Standard
Description
Includes
500μL (50 applications of 10μL each) provided in a plastic vial Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red
The SeeBlue™ Plus2 Pre-Stained Standard contains 10 proteins: 8 blue-dyed and 2 with contrasting colors, providing easier band identification. It can be used with NuPAGE™, tris-glycine, and tricine gels.
- 10 polypeptides resolved into 8 blue and 2 colored bands in the range of 4 to 250kDa (depending upon the buffer system)
- Immediate, unambiguous band identification
- A ready-to-use format—no mixing, heating, or reducing required
- Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels
This standard is pre-reduced and does not contain reducing reagents in the loading buffer.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | 500 μL (50 applications of 10 μL each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red. Store at 4°C. |
| Detection Method | Colorimetric |
| Number of Markers | 10 |
| Ready to Load | Yes |
| Size Range | 3 to 200 kDa |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels |
| Molecular Weight (g/mol) | 198, 98, 62, 49, 38, 28, 17, 14, 6, 3 kDa |
| Quantity | 500 μL |
| Shipping Condition | Wet Ice |
| Product Line | SeeBlue |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.