Learn More
American Educational Products Seed Germination and Plant Growth Kit
Shop All American Educational Products LLC Products

Description
Includes
- Seed packets
- Clear plastic germination chambers and growth domes
- Magnifying glass
- Teacher's guide
- Reproducible student worksheets
Germination chambers give students a clear view of the changes in their seeds (monocots and dicots provided), while growth domes give plants enough headroom to grow to transplantable size.
Order Info
Grades 3 to 8. Class size: 20 students.
Specifications
Specifications
| Grade | 3 to 8 |
| For Use With (Application) | Study the clear view of root and stem development |
| Includes | 4 packets of seeds, 5 germination chambers, growth domes, magnifying glass, teacher's guide and reproducible student worksheet |
| Class Size | 30 Students |
| Product Type | Seed Germination/Plant Growth Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.