Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Seipin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113923 Supplier Invitrogen™ Supplier No. PA5113923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEESTPQSDV TEDGESPEDP SGTEGQLSEE EKPDQQPLSG EEELEPEAS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 86%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Seipin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BSCL2
Gene Accession No. Q96G97
Gene Alias 2900097C17Rik; AI046355; Berardinelli-Seip congenital lipodystrophy 2 (seipin); Berardinelli-Seip congenital lipodystrophy 2 homolog (seipin); Bernardinelli-Seip congenital lipodystrophy 2 homolog; Bernardinelli-Seip congenital lipodystrophy type 2 protein; Bernardinelli-Seip congenital lipodystrophy type 2 protein homolog; bscl2; bscl2 {ECO:0000250; BSCL2, seipin lipid droplet biogenesis associated; G protein gamma 3 linked; Gng3lg; HMN5; MNCb-2630; PELD; seipin; SPG17; UniProtKB:Q96G97}
Gene Symbols BSCL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human BSCL2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26580
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.