Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP2K4 Monoclonal Antibody (2 E4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1098
Description
MA1-077 detects MAP2K4 in human samples and has been successfully used in Western blot and immunofluorescence applications.
MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
Specifications
MAP2K4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P45985 | |
MAP2K4 | |
Protein expressed in 293T cells transfected with human MAP2K4 expression vector. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
2 E4 | |
Unconjugated | |
MAP2K4 | |
C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein | |
Mouse | |
Protein A | |
RUO | |
6416 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction