Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP2K4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580790 Supplier Invitrogen™ Supplier No. PA580790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MKK4/MEK4/MAP2K4.

MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2K4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MAP2K4
Gene Accession No. P45985
Gene Alias C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein
Gene Symbols MAP2K4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human MKK4/MEK4/MAP2K4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.