Sekisui Diagnostics OSOM™ Trichomonas Rapid Test Kit Available on GSA/VA Contract for Federal Government customers only.

Immunochromatographic assay that detects Trichomonas antigens directly from vaginal swabs

Manufacturer: Sekisui Diagnostics 182

 View more versions of this product

Catalog No. 22-071-072

  Check Availability
Add to Cart

    Specifications

    Product Type Positive control kit
    Quantity 25 Tests
    Storage Requirements Room temperature
    Clia Complexity Waived
    CPT Code 87808QW
    For Use With (Equipment) OSOM™ Trichomonas rapid test kit
    Test Time 10 min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit