Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEKISUI Diagnostics™ OSOM™ Ultra Strep A Test
Shop All SEKISUI Diagnostics Products
Click to view available options
Quantity:
25 Tests
50 Tests
Description
Includes
Controls
- Simple-to-use - get easy to read results in four easy steps and no need for additional equipment
- Greater sensitivity - antibody labeled color particles coated at two separate locations allows for greater sensitivity than conventional single label technology without sacrificing specificity
- Easy-to-read two-color results
- Simplify reagent addition - single reagent delivery system in 25-test kit eliminates the need to handle two separate reagents
- QC Inside™ - two extra tests included in the 50-test kit at no additional charge to run external quality control for more confidence in your results
- Made in the USA

Specifications
Specifications
Accuracy | 92.6% |
Content And Storage | Room temperature (15°C to 30°C, 59°F to 86°F) |
Control Sets | Yes, included |
CPT Code | 87880QW |
Clia Complexity | Waived |
Detectable Analytes | Streptococcal Antigen (Group A) |
DoA Calibrators | No |
Sample Type | Throat Swab |
Sensitivity | 96.4% (95% CI: 93.0% to 97.9%) |
Type | Rapid Test |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction