Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEL1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518943
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518943 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518943 Supplier Invitrogen™ Supplier No. PA518943
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

The protein encoded by this gene is part of a protein complex required for the retrotranslocation or dislocation of misfolded proteins from the endoplasmic reticulum lumen to the cytosol, where they are degraded by the proteasome in a ubiquitin-dependent manner. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEL1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SEL1L
Gene Accession No. Q9UBV2
Gene Alias AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075
Gene Symbols SEL1L
Host Species Goat
Immunogen Synthetic peptide sequence (TAYNSYKDGDYN) corresponding to the internal amino acids of SEL1L (aa 561-573).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6400
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.