Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEL1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555035
Description
Immunogen sequence: REDGRLWCAT TYDYKADEKW GFCETEEEAA KRRQMQEAEM MYQTGMKILN GSNKKSQKRE AYRYLQKAAS MNHTKALERV SYALLFGDYL PQNIQAAREM FEKLTEEGSP KGQTALGFLY ASGLGVNSSQ AKALVYY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
The protein encoded by this gene is part of a protein complex required for the retrotranslocation or dislocation of misfolded proteins from the endoplasmic reticulum lumen to the cytosol, where they are degraded by the proteasome in a ubiquitin-dependent manner. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
SEL1L | |
Polyclonal | |
Unconjugated | |
SEL1L | |
AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6400 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBV2 | |
SEL1L | |
Recombinant protein corresponding to Human SEL1L. Recombinant protein control fragment (Product #RP-89019). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction