Learn More
Invitrogen™ SEL1L Polyclonal Antibody

Description
Immunogen sequence: SSDEEGSQDE SLDSKTTLTS DESVKDHTTA GRVVAGQIFL DSEESELESS IQEEEDSLKS QEGESVTEDI SFLESPNPEN KDYEEPKKVR KPALTAIEGT AHGEPCHFPF LFLDKEYDEC TSDGREDGRL WCATTYDYKA DEKWGFCETE EEAAKRRQMQ EAEMMYQTG; Positive Samples: BxPC-3, HepG2, 293T, Mouse kidney, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass type I membrane protein.
Specifications
Specifications
| Antigen | SEL1L |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | SEL1L |
| Gene Accession No. | Q80Z70, Q9UBV2, Q9Z2G6 |
| Gene Alias | AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075 |
| Gene Symbols | SEL1L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.