Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEL1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109312
Description
Positive Control: Zebrafish.
The protein encoded by this gene is part of a protein complex required for the retrotranslocation or dislocation of misfolded proteins from the endoplasmic reticulum lumen to the cytosol, where they are degraded by the proteasome in a ubiquitin-dependent manner. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| SEL1L | |
| Polyclonal | |
| Unconjugated | |
| SEL1L | |
| AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 568615 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| SEL1L | |
| Recombinant protein within zebrafish SEL1L aa 109-281. | |
| 100 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction