Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEL1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563659 Supplier Invitrogen™ Supplier No. PA563659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGSAGGNMMS QMILGYRYLS GINVLQNCEV ALSYYKKVAD YIADTFEKSE GVPVEKVRLT ERPENLSSNS EILDWDIYQY YKFLAERGDV QIQVSL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

SEL1L2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEL1L2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEL1L2
Gene Accession No. Q5TEA6
Gene Alias C20orf50; Gm118; Protein sel-1 homolog 2; RGD1311278; sel-1 suppressor of lin-12-like 2; sel-1 suppressor of lin-12-like 2 (C. elegans); Sel1l2; sel-1L2; SEL1L2 adaptor subunit of ERAD E3 ligase; SEL1L2 ERAD E3 ligase adaptor subunit; SEL1L2, ERAD E3 ligase adaptor subunit; suppressor of lin-12-like protein 2
Gene Symbols SEL1L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEL1L2. Recombinant protein control fragment (Product #RP-99325).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.