Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEL1L3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297792MFV500
Description
SEL1L3 Polyclonal antibody specifically detects SEL1L3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SEL1L3 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DKFZp781J1697, FLJ21629, KIAA0746FLJ41299, protein sel-1 homolog 3, sel-1 suppressor of lin-12-like 3 (C. elegans), Sel-1L3, Suppressor of lin-12-like protein 3 | |
Produced in rabbits immunized with E. coli-derived Human SEL1L3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
23231 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction