Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEL1L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559042 Supplier Invitrogen™ Supplier No. PA559042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMNLGYKHYQ GIDNYPLDWE LSYAYYSNIA TKTPLDQHTL QGDQAYVETI RLKDDEILKV QTKEDGDVFM WLKHEATRGN AAAQQRLAQM LFWGQQGVAK NPEAA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

SEL1L3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEL1L3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEL1L3
Gene Accession No. Q68CR1
Gene Alias KIAA0746; Protein sel-1 homolog 3; sel-1 suppressor of lin-12-like 3; SEL1L family member 3; SEL1L3; Sel-1L3; Suppressor of lin-12-like protein 3
Gene Symbols SEL1L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEL1L3. Recombinant protein control fragment (Product #RP-98346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.