Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEL1L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531795
Description
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (83%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SEL1L3 is a protein coding gene.
Specifications
SEL1L3 | |
Polyclonal | |
Unconjugated | |
SEL1L3 | |
KIAA0746; Protein sel-1 homolog 3; sel-1 suppressor of lin-12-like 3; SEL1L family member 3; SEL1L3; Sel-1L3; Suppressor of lin-12-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.92 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q68CR1 | |
SEL1L3 | |
Recombinant protein encompassing a sequence within the N-terminus region of human SEL1L3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction