Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Bis(2-ethylhexyl) sebacate, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111012804 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mL
25 mL

Bis(2-ethylhexyl) sebacate (BEHS) is a plasticizer widely used in preparing ion-selective electrodes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00009497
Synonym Di(2-ethylhexyl) sebacate; Sebacic acid di(2-ethylhexyl) ester; ′Dioctyl′ sebacate

Specifications

CAS 122-62-3
Density 0.914 g/mL (at 25°C)
Boiling Point 212°C (1 mmHg)
Flash Point >113°C
Linear Formula [-(CH2)4 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 25 mL
Formula Weight 426.67
Percent Purity ≥97.0% (GC); ≥97.0%
Grade Selectophore
Product Line Selectophore
RTECSNumber VS1000000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.