Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Selectophore™ Diisodecyl phthalate, ≥99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mL
50 mL
Specifications
Specifications
Density | 0.965 g/mL (at 20°C) |
CAS | 26761-40-0 |
Flash Point | 275°C |
Linear Formula | C28H46O4 |
Quantity | 50 mL |
Formula Weight | 446.66 |
Percent Purity | ≥99.0% (sum of isomers, GC); ≥99.0% |
Grade | Selectophore |
Product Line | Selectophore |
Safety and Handling
RTECSNumber | TI1270000 |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction