Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Selectophore™ Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥98.0% (based on Na, AAS), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Specifications
Specifications
CAS | 79060-88-1 |
Quantity | 500 mg |
Linear Formula | C32H12 BF24 Na |
Formula Weight | 886.20 |
Percent Purity | ≥98.0% (based on Na, AAS) |
Grade | Selectophore™ |
Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
Product Line | Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction