Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELENBP1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297662PCP
Description
SELENBP1 Polyclonal antibody specifically detects SELENBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SELENBP1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813 | |
Produced in rabbits immunized with E. coli-derived Human SELENBP1 fragment. | |
0.1 mL | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
8991 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction