Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELENBP1 Monoclonal Antibody (OTI3D4), TrueMAB™, OriGene
SDP

Catalog No. 502540163
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0163 30 μL
50-167-5333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0163 Supplier OriGene Supplier No. TA504702S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SELENBP1 Monoclonal antibody specifically detects SELENBP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SELENBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SELENBP1
Gene Accession No. P17563, Q13228
Gene Alias 56 kDa selenium-binding protein; epididymis secretory sperm binding protein Li 134P; FLJ13813; HEL-S-134P; hSBP; hSP56; Lp 56; Lp56; LPSB; Methanethiol oxidase; methanethiol oxidase; selenium-binding protein 1; MTO; SBP; SBP 1; SBP1; SBP56; Selen bp2; SELENBP 1; Selenbp 2; selenbp1; Selenbp2; selenium binding protein 1; selenium binding protein 2; selenium-binding protein 1; Selenium-binding protein 2; SELNBP1; similar to selenium binding protein 1; SP 56; SP56; zgc:65844
Host Species Mouse
Immunogen Full length human recombinant protein of human SELENB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20341, 8991
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.