Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SELENBP1 Monoclonal Antibody (OTI3H1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504701S


Description
SELENBP1 Monoclonal antibody specifically detects SELENBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
SELENBP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13228 | |
Mouse | |
Affinity chromatography | |
RUO | |
8991 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3H1 | |
Unconjugated | |
SELENBP1 | |
56 kDa selenium-binding protein; epididymis secretory sperm binding protein Li 134P; FLJ13813; HEL-S-134P; hSBP; hSP56; Lp 56; Lp56; LPSB; Methanethiol oxidase; methanethiol oxidase; selenium-binding protein 1; MTO; SBP; SBP 1; SBP1; SBP56; Selen bp2; SELENBP 1; Selenbp 2; selenbp1; Selenbp2; selenium binding protein 1; selenium binding protein 2; selenium-binding protein 1; Selenium-binding protein 2; SELNBP1; similar to selenium binding protein 1; SP 56; SP56; zgc:65844 | |
Full length human recombinant protein of human SELENB1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction