Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SELENBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543146 Supplier Invitrogen™ Supplier No. PA543146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MATKCGNCGP GYSTPLEAMK GPREEIVYLP CIYRNTGTEA PDYLATVDVD Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colorectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SELENBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SELENBP1
Gene Accession No. Q13228
Gene Alias 56 kDa selenium-binding protein; epididymis secretory sperm binding protein Li 134P; FLJ13813; HEL-S-134P; hSBP; hSP56; Lp 56; Lp56; LPSB; Methanethiol oxidase; methanethiol oxidase; selenium-binding protein 1; MTO; SBP; SBP 1; SBP1; SBP56; Selen bp2; SELENBP 1; Selenbp 2; selenbp1; Selenbp2; selenium binding protein 1; selenium binding protein 2; selenium-binding protein 1; Selenium-binding protein 2; SELNBP1; similar to selenium binding protein 1; SP 56; SP56; zgc:65844
Gene Symbols SELENBP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SELENBP1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8991
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.