Learn More
Selenic acid, 40% aq. soln., Thermo Scientific Chemicals
Description
Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 g |
Molecular Formula | K2O4Se |
Linear Formula | H2SeO4 |
UN Number | UN2922 |
Merck Index | 14,8429 |
Solubility Information | Miscible with sulfuric acid. Immiscible with ammonia. |
InChI Key | YAZJAPBTUDGMKO-UHFFFAOYSA-L |
SMILES | O[Se](=O)(=O)O |
Show More |
Safety and Handling
Hazard Category | H301+H331-H373 |
Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 231-979-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.