Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenic acid, 40% aq. soln.

Catalog No. p-7049704
Click to view available options
Quantity:
100 g
500 g

CAS: 7783-08-6 | K2O4Se | 221.16 g/mol

Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-08-6
MDL Number MFCD00012191
Synonym selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2
ChEBI CHEBI:18170

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 g
Molecular Formula K2O4Se
Linear Formula H2SeO4
UN Number UN2922
Merck Index 14,8429
Solubility Information Miscible with sulfuric acid. Immiscible with ammonia.
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
SMILES O[Se](=O)(=O)O
IUPAC Name selenic acid
Molecular Weight (g/mol) 221.16
PubChem CID 1089
Formula Weight 144.97 (Anhydrous)
Concentration or Composition (by Analyte or Components) 40% aq. soln.
Density 1.41 g/mL
Chemical Name or Material Selenic acid
Show More Show Less
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 231-979-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.