Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium, AAS standard solution, Specpure™ Se 1000μg/mL

Catalog No. p-4408592
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-49-2, 7697-37-2, 7732-18-5
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Vapor Pressure 17mm Hg at 20°C
Assay Percent Range Se 1000μg/mL
Physical Form Liquid
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Selenium: 0.1%; Water: 94.9%
Chemical Name or Material Selenium
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.