Alfa Aesar™ Selenium dichloride oxide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 68108AC

Quantity 25mL

 View more versions of this product

Catalog No. AA68108AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-23-3
    Molecular Formula SeOCl2
    Formula Weight 165.87
    MDL Number MFCD00016327
    UN Number UN2879
    Merck Index 14,8435
    Synonym Selenium oxychloride
    Chemical Name or Material Selenium dichloride oxide

    Specifications

    Formula Weight 165.87
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.651
    CAS 7791-23-3
    Color Yellow
    Sensitivity Moisture Sensitive
    Boiling Point 176.4°C
    Melting Point 8.5°C
    Solubility Information Decomposes in water forming HCl and H2SeO3
    Health Hazard 1 Danger
    Density 2.42g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: SELENIUM OXYCHLORIDE
    TSCA TSCA
    Molecular Formula SeOCl2
    MDL Number MFCD00016327
    Synonym Selenium oxychloride
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Physical Form Liquid
    UN Number UN2879
    Merck Index 14,8435
    Quantity 25mL
    Chemical Name or Material Selenium dichloride oxide
    Assay 99%, (metals basis)

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit