Selenium(IV) oxide, 99.4% (Metals Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1235822

Quantity 100g

 View more versions of this product

Catalog No. AA1235822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-08-4
    Physical Form Powder or Lump
    Assay Percent Range 99.4% (Metals Basis)
    Molecular Formula O2Se
    Formula Weight 110.96
    MDL Number MFCD00003562
    Synonym Selenium dioxide
    Chemical Name or Material Selenium(IV) oxide

    Specifications

    Physical Form Powder or Lump
    Formula Weight 110.96
    Quantity 100g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: II
    Density 3.95g/cm3 at 20°C
    EINECS Number 231-194-7
    Odor Odorless
    Assay Percent Range 99.4% (Metals Basis)
    Molecular Formula O2Se
    MDL Number MFCD00003562
    CAS 7446-08-4
    Synonym Selenium dioxide
    Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Health Hazard 2 When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
    Chemical Name or Material Selenium(IV) oxide

    When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

    H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit